missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AlphaProTech Critical Cover™ GenPro™ Coveralls
Soft polypropylene coveralls are ideal for non-hazardous environments. Lightweight, durable material lets the wearer stay clean and comfortable all day while providing a limited barrier to particulates and contamination.
$190.20 - $235.57
Specifications
| Product Type | Cleanroom Coveralls |
|---|---|
| Material | Polypropylene |
| Color | White |
| Disposable | Single-use |
| Ankle Style | Elastic |
| Catalog Number | Mfr. No. | Size | Size (English) Chest | Length (English) Sleeve | Size (English) Inseam | Price | Quantity | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Size (English) Chest | Length (English) Sleeve | Size (English) Inseam | Price | Quantity | ||
|
19-169-4005
|
AlphaProTech
CV140322 |
Medium | 24.25 in. | 34 in. | 28.5 in. |
Case of 25 Each for $190.20
|
|
||
|
19-169-4006
|
AlphaProTech
CV140323 |
Large | 25 in. | 34 in. | 29.5 in. |
Case of 25 Each for $190.20
|
|
||
|
19-169-4007
|
AlphaProTech
CV140324 |
X-Large | 27 in. | 36.5 in. | 30 in. |
Case of 25 Each for $190.20
|
|
||
|
19-169-4008
|
AlphaProTech
CV140325 |
2X-Large | 29.75 in. | 37 in. | 31 in. |
Case of 25 Each for $196.05
|
|
||
|
19-169-4009
|
AlphaProTech
CV140326 |
3X-Large | 32 in. | 38 in. | 32 in. |
Case of 25 Each for $235.57
|
|
||
|
19-165-640
|
AlphaProTech
CV140327 |
4X-Large | 32.5 in. | 41 in. | 32.5 in. |
Case of 25 Each for $221.22
|
|
||
|
19-169-4010
|
AlphaProTech
CV140328 |
5X-Large | 33.5 in. | 42 in. | 33 in. |
Case of 25 Each for $213.64
|
|
||
Description
- Lightweight, durable material
- Elastic back
- Elastic wrist, ankle and back
- Serged seams
- No pockets
Specifications
| Cleanroom Coveralls | |
| White | |
| Elastic | |
| Serged | |
| Elastic | |
| Yes | |
| GenPro™ |
| Polypropylene | |
| Single-use | |
| Elastic | |
| Non-sterile | |
| Elastic wrist, ankle and back | |
| Provides limited barrier to particulates and contamination in non-hazardous environments |