missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AlphaProTech Critical Cover™ GenPro™ Coveralls
Soft polypropylene coveralls are ideal for non-hazardous environments. Lightweight, durable material lets the wearer stay clean and comfortable all day while providing a limited barrier to particulates and contamination.
Supplier: AlphaProTech CV140326
Description
- Lightweight, durable material
- Elastic back
- Elastic wrist, ankle and back
- Serged seams
- No pockets
Specifications
| Cleanroom Coveralls | |
| Polypropylene | |
| Single-use | |
| Elastic | |
| Non-sterile | |
| Elastic wrist, ankle and back | |
| 32 in. | |
| 81.28 cm | |
| Provides limited barrier to particulates and contamination in non-hazardous environments | |
| 96.52 cm |
| 3X-Large | |
| White | |
| Elastic | |
| Serged | |
| Elastic | |
| Yes | |
| 32 in. | |
| 81.28 cm | |
| 38 in. | |
| GenPro™ |