missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha Tubulin Antibody (YL1/2), HRP, R&D Systems™
Description
alpha Tubulin Monoclonal specifically detects alpha Tubulin in Avian, C. elegans, Drosophila, Human, Invertebrate, Mammalia, Monkey, Mouse, Primate, Rat, Yeast samples. It is validated for ELISA, Flow Cytometry, Functional Assay, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Radioimmune Assays (RIA), Western Blot.
Specifications
Specifications
| Antigen | alpha Tubulin |
| Applications | ELISA, Flow Cytometry, Functional Assay, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Radioimmune Assays (RIA), Western Blot |
| Classification | Monoclonal |
| Clone | YL1/2 |
| Conjugate | HRP |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen |
| Formulation | PBS with No Preservative |
| Gene Alias | Alpha-tubulin 3, B-ALPHA-1, FLJ25113, hum-a-tub1, hum-a-tub2, LIS3, TUBA3, tubulin alpha-1A chain, Tubulin alpha-3 chain, Tubulin B-alpha-1, tubulin, alpha 1a, tubulin, alpha 3, tubulin, alpha, brain-specific |
| Gene Symbols | TUBA1A |
| Host Species | Rat |
| Show More |