missing translation for 'onlineSavingsMsg'
Learn More

alpha-(Trifluoromethyl)benzyl Alcohol 98.0+%, TCI America™

Catalog No. T17925G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17925G 5 g
T179225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T17925G Supplier TCI America Supplier No. T17925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 340-05-6
Molecular Formula C8H7F3O
Molecular Weight (g/mol) 176.14
MDL Number MFCD00004498
InChI Key VNOMEAQPOMDWSR-UHFFFAOYNA-N
Synonym 1-phenyl-2,2,2-trifluoroethanol, alpha-trifluoromethyl benzyl alcohol, 2,2,2-trifluoro-1-phenylethan-1-ol, alpha-trifluoromethyl benzyl?alcohol, alpha-trifluoromethyl benzenemethanol, 2,2,2-trifluoro-1-phenyl-ethanol, benzenemethanol, alpha-trifluoromethyl, 2,2,2-tris fluoranyl-1-phenyl-ethanol, 2,2,2-trifluorophenylethanol
PubChem CID 95556
IUPAC Name 2,2,2-trifluoro-1-phenylethan-1-ol
SMILES OC(C1=CC=CC=C1)C(F)(F)F

Specifications

Boiling Point 88°C
Quantity 5 g
Formula Weight 176.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material alpha-(Trifluoromethyl)benzyl Alcohol