accessibility menu, dialog, popup

UserName

OHFFF

CAS RN 340-05-6

IUPAC Name: 2,2,2-trifluoro-1-phenylethan-1-ol
Synonyms: 2,2,2-trifluoro-1-phenylethanol
Molecular Weight (g/mol): 176.14
Molecular Formula: C8H7F3O
InChi Key: VNOMEAQPOMDWSR-UHFFFAOYNA-N
SMILES: OC(C1=CC=CC=C1)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results