missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Thioglycerol 98.0+%, TCI America™
$44.73 - $177.08
Chemical Identifiers
| CAS | 96-27-5 |
|---|---|
| Molecular Formula | C3H8O2S |
| Molecular Weight (g/mol) | 108.16 |
| MDL Number | MFCD00004879 |
| InChI Key | PJUIMOJAAPLTRJ-UHFFFAOYNA-N |
| Synonym | 3-mercapto-1,2-propanediol, monothioglycerol, 1-thioglycerol, thioglycerol, thioglycerine, thioglycerin, thiovanol, 3-mercaptopropane-1,2-diol, alpha-thioglycerol, glycerol-1-thiol |
| PubChem CID | 7291 |
| ChEBI | CHEBI:74537 |
| IUPAC Name | 3-sulfanylpropane-1,2-diol |
| SMILES | OCC(O)CS |
Chemical Identifiers
| 96-27-5 | |
| 108.16 | |
| PJUIMOJAAPLTRJ-UHFFFAOYNA-N | |
| 7291 | |
| 3-sulfanylpropane-1,2-diol |
| C3H8O2S | |
| MFCD00004879 | |
| 3-mercapto-1,2-propanediol, monothioglycerol, 1-thioglycerol, thioglycerol, thioglycerine, thioglycerin, thiovanol, 3-mercaptopropane-1,2-diol, alpha-thioglycerol, glycerol-1-thiol | |
| CHEBI:74537 | |
| OCC(O)CS |
Specifications
| 96-27-5 | |
| C3H8O2S | |
| 1 g | |
| 3-mercapto-1,2-propanediol, monothioglycerol, 1-thioglycerol, thioglycerol, thioglycerine, thioglycerin, thiovanol, 3-mercaptopropane-1,2-diol, alpha-thioglycerol, glycerol-1-thiol | |
| OCC(O)CS | |
| 108.16 | |
| CHEBI:74537 | |
| ≥98.0% (GC,T) | |
| alpha-Thioglycerol [Matrix for FABMS and liquid SIMS] |
| Colorless | |
| MFCD00004879 | |
| 2810 | |
| PJUIMOJAAPLTRJ-UHFFFAOYNA-N | |
| 3-sulfanylpropane-1,2-diol | |
| 7291 | |
| 108.16 | |
| Liquid |
Safety and Handling
EINECSNumber : (9)-0805
RTECSNumber : TY8140000
TSCA : Yes