missing translation for 'onlineSavingsMsg'
Learn More

alpha-Tetralone 98.0+%, TCI America™

Catalog No. T013425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T013425G 25 g
T0134500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T013425G Supplier TCI America Supplier No. T013425G

Chemical Identifiers

CAS 529-34-0
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.19
MDL Number MFCD00001688
InChI Key XHLHPRDBBAGVEG-UHFFFAOYSA-N
Synonym alpha-tetralone, 1-tetralone, 3,4-dihydronaphthalen-1 2h-one, 1,2,3,4-tetrahydronaphthalen-1-one, 3,4-dihydro-1 2h-naphthalenone, tetralone, a-tetralone, 1-oxotetralin, 1 2h-naphthalenone, 3,4-dihydro, .alpha.-tetralone
PubChem CID 10724
IUPAC Name 1,2,3,4-tetrahydronaphthalen-1-one
SMILES O=C1CCCC2=CC=CC=C12

Specifications

Color Yellow
Boiling Point 257°C
Quantity 25 g
Formula Weight 146.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material alpha-Tetralone
EINECSNumber (9)-0831
RTECSNumber QK4375000
TSCA Yes