missing translation for 'onlineSavingsMsg'
Learn More

alpha-Terpineol, Practical, 80-90%, Spectrum™ Chemical

Catalog No. 18600446
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
Packaging:
Steel Can
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600446 500 mL Steel Can
18600447 4 L Steel Can
18600448 20 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18600446 Supplier Spectrum Chemical Mfg Cor Supplier No. TE107500MLGL
Get Up to $500 in Chemicals with Purchase
View all promotions

TE107, 98-55-5, C10H18O

Spectrum™ Chemical alpha-Terpineol, Practical , is a naturally occurring monoterpene alcohol often isolated from pine oil. The Alpha isomer is one of three isomers, and is generally the most common constituent. It has a pleasenat lilac odor and for this reason is often used in perfumes, cosmetics and flavoring agents.

Chemical Identifiers

CAS 98-55-5
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
InChI Key WUOACPNHFRMFPN-VIFPVBQESA-N
IUPAC Name 2-[(1R)-4-methylcyclohex-3-en-1-yl]propan-2-ol
SMILES CC1=CC[C@@H](CC1)C(C)(C)O

Specifications

CAS Max % 100%
Packaging Steel Can
Quantity 500 mL
Percent Purity 80 to 90%
Grade Practical
Physical Form Liquid
Chemical Name or Material alpha-Terpineol