missing translation for 'onlineSavingsMsg'
Learn More

alpha-Terpinene, 90+%

CAS: 99-86-5 | C10H16 | 136.24 g/mol

$61.10 - $167.30

Chemical Identifiers

CAS 99-86-5
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00001534
InChI Key YHQGMYUVUMAZJR-UHFFFAOYSA-N
Synonym alpha-terpinene, p-mentha-1,3-diene, terpilene, 1-isopropyl-4-methyl-1,3-cyclohexadiene, 1,3-cyclohexadiene, 1-methyl-4-1-methylethyl, alpha-terpinen, 1-methyl-4-1-methylethyl-1,3-cyclohexadiene, 1-methyl-4-isopropylcyclohexadiene-1,3, .alpha.-terpinen, .alpha.-terpinene
PubChem CID 7462
ChEBI CHEBI:10334
IUPAC Name 1-methyl-4-propan-2-ylcyclohexa-1,3-diene
SMILES CC(C)C1=CC=C(C)CC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ62871AE
View Documents Promotion Details
N/A 100 mL
Each for $61.10
Only null left
 
AAJ62871AP
View Documents Promotion Details
N/A 500 mL
Each for $167.30
Only null left
 
Description

Description

It can be used as a chemical fragrance essential oil and spices. It is used as a flavoring and fragrance chemical used in the personal care and cosmetic and food industries. Also used in the pharmaceutical, electronics, food processing, semi-conductor manufacturing industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used as a chemical fragrance essential oil and spices. It is used as a flavoring and fragrance chemical used in the personal care and cosmetic and food industries. Also used in the pharmaceutical, electronics, food processing, semi-conductor manufacturing industries.

Solubility
Soluble in ether, alcohol, and ethanol (100 mg/ml). Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature: -20°C. Store under inert gas. Air sensitive. Keep away from strong acids, strong bases.
Specifications

Chemical Identifiers

99-86-5
136.24
YHQGMYUVUMAZJR-UHFFFAOYSA-N
7462
1-methyl-4-propan-2-ylcyclohexa-1,3-diene
C10H16
MFCD00001534
alpha-terpinene, p-mentha-1,3-diene, terpilene, 1-isopropyl-4-methyl-1,3-cyclohexadiene, 1,3-cyclohexadiene, 1-methyl-4-1-methylethyl, alpha-terpinen, 1-methyl-4-1-methylethyl-1,3-cyclohexadiene, 1-methyl-4-isopropylcyclohexadiene-1,3, .alpha.-terpinen, .alpha.-terpinene
CHEBI:10334
CC(C)C1=CC=C(C)CC1

Specifications

99-86-5
0.837
50°C (122°F)
1.478
UN2319
14,9170
Soluble in ether,alcohol,and ethanol (100mg/ml). Insoluble in water.
CC(C)C1=CC=C(C)CC1
136.24
CHEBI:10334
≥90%
alpha-Terpinene
Colorless
173°C to 175°C
C10H16
MFCD00001534
1853379
alpha-terpinene, p-mentha-1,3-diene, terpilene, 1-isopropyl-4-methyl-1,3-cyclohexadiene, 1,3-cyclohexadiene, 1-methyl-4-1-methylethyl, alpha-terpinen, 1-methyl-4-1-methylethyl-1,3-cyclohexadiene, 1-methyl-4-isopropylcyclohexadiene-1,3, .alpha.-terpinen, .alpha.-terpinene
YHQGMYUVUMAZJR-UHFFFAOYSA-N
1-methyl-4-propan-2-ylcyclohexa-1,3-diene
7462
136.23
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H302-H315-H319-H335
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P272-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P331-P333+P313-P363-P370+P378q-P501c

H226-H302-H304-H317-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: TERPENE HYDROCARBONS, N.O.S.

EINECSNumber : 202-795-1

RTECSNumber : OS8060000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only