missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Alpha-Smooth Muscle Actin Recombinant Rabbit Monoclonal Antibody (23GB600)
GREENER_CHOICE

Catalog No. MA554092
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA554092 100 μL
MA554093 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA554092 Supplier Invitrogen™ Supplier No. MA554092
Only null left

Rabbit Recombinant Monoclonal Antibody

Alpha-Smooth Muscle Actin Recombinant Monoclonal Antibody for Flow, ICC/IF, IHC (P), Western Blot

Smooth Muscle Actin belongs to the actin family of proteins, which are highly conserved proteins that play a role in cell motility, structure and integrity. Alpha, beta and gamma actin isoforms have been identified, with alpha actin being a major constituent of the contractile apparatus, while beta and gamma actins are involved in the regulation of cell motility. In particular, smooth muscle actin is an alpha actin that is found in skeletal muscle. Actin exists as a ubiquitous protein involved with filament formation that make up large portions of the cytoskeleton. Actin filaments interact with myosin to assist in muscle contraction as well as aiding in cell motility and cytokinesis. Smooth muscle actin is found on smooth muscle vessel walls, gut wall, myometrium, myoepithelial cells in breast and salivary glands. Defects in the smooth muscle actin gene cause aortic aneurysm familial thoracic type 6. Actin isoforms differ slightly in their N-terminus and the sequences of each are perfectly conserved in higher vertebrates. Alpha-smooth muscle actin is abundant in vascular and visceral smooth muscle cells. In addition, it has also been shown that smooth muscle actin appear in stress fibers of fibroblastic cells during pathological situations involving contractile phenomena such as wound healing and fibrocontractive diseases. Multiple alternatively spliced variants of smooth muscle actin have been identified.

Specifications

Antigen Alpha-Smooth Muscle Actin
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB600
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ACTA2
Gene Accession No. P62736, P62737, P62738
Gene Alias 0610041G09Rik; AAT6; ACT-4; acta2; acta2.L; acta2.S; actin; Actin Alpha 2 Smooth Muscle Aorta; actin alpha 2, smooth muscle; Actin Vascular Smooth Muscle; actin, alpha 2, smooth muscle, aorta; actin, alpha 2, smooth muscle, aorta L homeolog; actin, alpha 2, smooth muscle, aorta S homeolog; actin, alpha, vascular smooth muscle; actin, aortic smooth muscle; Actin, aortic smooth muscle, intermediate form; ACTSA; ACTVS; alpha actin 2; Alpha Cardiac Actin; alpha smooth muscle actin; alpha-actin; Alpha-actin-2; alpha-cardiac actin; alphaSMA; alpha-SM-actin; alpha-smooth muscle actin; Aortic Smooth Muscle; asma; a-sma; cell growth-inhibiting gene 46 protein; GIG46; Growth Inhibiting Gene 46; hypothetical protein LOC515610; LOW QUALITY PROTEIN: actin, aortic smooth muscle; mymy5; SM actin; sma; SMactin; sm-actin; SMalphaA; smooth muscle; smooth muscle alpha-actin; vascular smooth muscle alpha-actin; wu:fb63d03; XELAEV_18034370mg
Gene Symbols ACTA2
Host Species Rabbit
Immunogen A synthetic peptide of human alpha smooth muscle Actin (1-54AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11475, 59, 81633
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less