missing translation for 'onlineSavingsMsg'
Learn More

alpha-Pinene Oxide 95.0+%, TCI America™

Catalog No. p-7135098
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P136225ML 25 mL
P1362250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P136225ML Supplier TCI America Supplier No. P136225ML
Only null left

Chemical Identifiers

CAS 1686-14-2
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.237
MDL Number MFCD00066955
InChI Key NQFUSWIGRKFAHK-UHFFFAOYSA-N
Synonym 2,3-Epoxypinane
PubChem CID 91508
ChEBI CHEBI:29060
SMILES CC1(C2CC1C3(C(C2)O3)C)C

Specifications

Color Yellow
Quantity 25 mL
UN Number 2810
Formula Weight 152.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material alpha-Pinene Oxide
EINECSNumber (4)-0616
RTECSNumber TK4565000
TSCA Yes