CAS RN 1686-14-2
IUPAC Name:
2,7,7-trimethyl-3-oxatricyclo[4.1.1.0²,⁴]octane
Synonyms:
α-pinene oxide 2-pinene oxide α-pinene epoxide
Molecular Weight (g/mol):
152.24
Molecular Formula:
C10H16O
InChi Key:
NQFUSWIGRKFAHK-UHFFFAOYNA-N
SMILES:
CC12OC1CC1CC2C1(C)C
1
–
1
of
1
results
1
alpha-Pinene Oxide 95.0+%, TCI America™
CAS: 1686-14-2 Molecular Formula: C10H16O Molecular Weight (g/mol): 152.237 MDL Number: MFCD00066955 InChI Key: NQFUSWIGRKFAHK-UHFFFAOYSA-N Synonym: 2,3-Epoxypinane PubChem CID: 91508 ChEBI: CHEBI:29060 SMILES: CC1(C2CC1C3(C(C2)O3)C)C