missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Pinene Oxide 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 25 mL |
| UN Number | 2810 |
| Formula Weight | 152.24 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | alpha-Pinene Oxide |
Safety and Handling
| EINECSNumber | (4)-0616 |
| RTECSNumber | TK4565000 |
| TSCA | Yes |