Learn More
(-)-alpha-Pinene, 98%, cont. variable amounts of enantiomer
CAS: 7785-26-4 | C10H16 | 136.238 g/mol
$126.74
Chemical Identifiers
| CAS | 7785-26-4 |
|---|---|
| Molecular Formula | C10H16 |
| Molecular Weight (g/mol) | 136.238 |
| MDL Number | MFCD00064145 |
| InChI Key | GRWFGVWFFZKLTI-IUCAKERBSA-N |
| Synonym | --alpha-pinene, 1s,5s-2,6,6-trimethylbicyclo 3.1.1 hept-2-ene, 1s---alpha-pinene, unii-tzr3gm95pr, dl-alpha-pinene, alpha-pinene,-, 1s,5s-alpha-pinene, tzr3gm95pr, 1s,5s-2-pinene, bicyclo 3.1.1 hept-2-ene, 2,6,6-trimethyl-, 1s,5s |
| PubChem CID | 440968 |
| ChEBI | CHEBI:28660 |
| IUPAC Name | (1S,5S)-4,6,6-trimethylbicyclo[3.1.1]hept-3-ene |
| SMILES | CC1=CCC2CC1C2(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAL0475930
|
N/A | 250 g |
Each for $126.74
|
|
|||||
Description
(-)-alpha-Pinene, 98%, cont. variable amounts of enantiomer, is used in food and beverage (incl. additives, contaminants), fragrance / essential oil, nutraceutical. It is also used as a anti- infective agent. It is used as a component of pine resin and a useful chiral synthetic intermediate. Alpha-pinene is the most widely encountered terpenoid in nature and is highly repellant to insects.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(-)-alpha-Pinene, 98%, cont. variable amounts of enantiomer, is used in food and beverage (incl. additives, contaminants), fragrance / essential oil, nutraceutical. It is also used as a anti- infective agent. It is used as a component of pine resin and a useful chiral synthetic intermediate. Alpha-pinene is the most widely encountered terpenoid in nature and is highly repellant to insects.
Solubility
Soluble in chloroform, ether, alcohol, and methanol. Insoluble in water.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
Chemical Identifiers
| 7785-26-4 | |
| 136.238 | |
| GRWFGVWFFZKLTI-IUCAKERBSA-N | |
| 440968 | |
| (1S,5S)-4,6,6-trimethylbicyclo[3.1.1]hept-3-ene |
| C10H16 | |
| MFCD00064145 | |
| --alpha-pinene, 1s,5s-2,6,6-trimethylbicyclo 3.1.1 hept-2-ene, 1s---alpha-pinene, unii-tzr3gm95pr, dl-alpha-pinene, alpha-pinene,-, 1s,5s-alpha-pinene, tzr3gm95pr, 1s,5s-2-pinene, bicyclo 3.1.1 hept-2-ene, 2,6,6-trimethyl-, 1s,5s | |
| CHEBI:28660 | |
| CC1=CCC2CC1C2(C)C |
Specifications
| 7785-26-4 | |
| 0.856 | |
| 32°C (89°F) | |
| 1.465 | |
| UN2368 | |
| 14,7445 | |
| Soluble in chloroform,ether,alcohol,and methanol. Insoluble in water. | |
| −42° (Neat) | |
| (1S,5S)-4,6,6-trimethylbicyclo[3.1.1]hept-3-ene | |
| 440968 | |
| 136.24 | |
| cont. variable amounts of enantiomer |
| -64°C | |
| 154°C to 156°C | |
| C10H16 | |
| MFCD00064145 | |
| 1903790 | |
| --alpha-pinene, 1s,5s-2,6,6-trimethylbicyclo 3.1.1 hept-2-ene, 1s---alpha-pinene, unii-tzr3gm95pr, dl-alpha-pinene, alpha-pinene,-, 1s,5s-alpha-pinene, tzr3gm95pr, 1s,5s-2-pinene, bicyclo 3.1.1 hept-2-ene, 2,6,6-trimethyl-, 1s,5s | |
| GRWFGVWFFZKLTI-IUCAKERBSA-N | |
| CC1=CCC2CC1C2(C)C | |
| 136.238 | |
| CHEBI:28660 | |
| 98% | |
| (-)-alpha-Pinene |
Safety and Handling
GHS H Statement
H304-H226-H302-H312-H332-H315-H319-H317
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
P210-P233-P240-P241-P242-P243-P261-P264b-P270-P272-P280-P301+P310-P303+P361+P353-P305+P351+P338-P330-P331-P333+P313-P363-P370+P378q-P501c
H226-H302-H304-H315-H317-H319
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALPHA-PINENE
EINECSNumber : 232-077-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only