missing translation for 'onlineSavingsMsg'
Learn More

(-)-alpha-Pinene, 98%, cont. variable amounts of enantiomer

Catalog No. AAL0475930
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0475930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0475930 Supplier Thermo Scientific Chemicals Supplier No. L0475930
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7785-26-4 | C10H16 | 136.238 g/mol

(-)-alpha-Pinene, 98%, cont. variable amounts of enantiomer, is used in food and beverage (incl. additives, contaminants), fragrance / essential oil, nutraceutical. It is also used as a anti- infective agent. It is used as a component of pine resin and a useful chiral synthetic intermediate. Alpha-pinene is the most widely encountered terpenoid in nature and is highly repellant to insects.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(-)-alpha-Pinene, 98%, cont. variable amounts of enantiomer, is used in food and beverage (incl. additives, contaminants), fragrance / essential oil, nutraceutical. It is also used as a anti- infective agent. It is used as a component of pine resin and a useful chiral synthetic intermediate. Alpha-pinene is the most widely encountered terpenoid in nature and is highly repellant to insects.

Solubility
Soluble in chloroform, ether, alcohol, and methanol. Insoluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.

Chemical Identifiers

CAS 7785-26-4
Molecular Formula C10H16
Molecular Weight (g/mol) 136.238
MDL Number MFCD00064145
InChI Key GRWFGVWFFZKLTI-IUCAKERBSA-N
Synonym --alpha-pinene, 1s,5s-2,6,6-trimethylbicyclo 3.1.1 hept-2-ene, 1s---alpha-pinene, unii-tzr3gm95pr, dl-alpha-pinene, alpha-pinene,-, 1s,5s-alpha-pinene, tzr3gm95pr, 1s,5s-2-pinene, bicyclo 3.1.1 hept-2-ene, 2,6,6-trimethyl-, 1s,5s
PubChem CID 440968
ChEBI CHEBI:28660
IUPAC Name (1S,5S)-4,6,6-trimethylbicyclo[3.1.1]hept-3-ene
SMILES CC1=CCC2CC1C2(C)C

Specifications

Melting Point -64°C
Density 0.856
Boiling Point 154°C to 156°C
Flash Point 32°C (89°F)
Refractive Index 1.465
Quantity 250 g
UN Number UN2368
Beilstein 1903790
Merck Index 14,7445
Solubility Information Soluble in chloroform,ether,alcohol,and methanol. Insoluble in water.
Optical Rotation −42° (Neat)
Formula Weight 136.24
Percent Purity 98%
Assay cont. variable amounts of enantiomer
Chemical Name or Material (-)-alpha-Pinene
Show More Show Less
Hazard Category H226-H302-H304-H315-H317-H319
Hazard Statement GHS H Statement
H304-H226-H302-H312-H332-H315-H319-H317
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P272-P280-P301+P310-P303+P361+P353-P305+P351+P338-P330-P331-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALPHA-PINENE
EINECSNumber 232-077-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only