missing translation for 'onlineSavingsMsg'
Learn More

alpha-(p-Toluenesulfonyl)benzyl Isocyanide 98.0+%, TCI America™

Catalog No. T31571G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31571G 1 g
T31575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T31571G Supplier TCI America Supplier No. T31571G

Chemical Identifiers

CAS 36635-66-2
Molecular Formula C15H13NO2S
Molecular Weight (g/mol) 271.334
MDL Number MFCD04114776
InChI Key KIJCBVOPJSHLBI-UHFFFAOYSA-N
Synonym alpha-Tosylbenzyl Isocyanide
PubChem CID 10967613
IUPAC Name 1-[isocyano(phenyl)methyl]sulfonyl-4-methylbenzene
SMILES CC1=CC=C(C=C1)S(=O)(=O)C(C2=CC=CC=C2)[N+]#[C-]

Specifications

Melting Point 106°C
Quantity 1 g
Formula Weight 271.33
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material alpha-(p-Toluenesulfonyl)benzyl Isocyanide