missing translation for 'onlineSavingsMsg'
Learn More

alpha-Naphtholbenzein

Catalog No. p-7023065
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1612506 5 g
AAA1612514 25 g
AAA1612522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1612506 Supplier Thermo Scientific Chemicals Supplier No. A1612506
Only null left

CAS: 145-50-6 | C27H18O2 | 374.439 g/mol

α-Naphtholbenzein is used as a pH indicator. (pH 0.0-0.8/ pH 8.2-10.0).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
α-Naphtholbenzein is used as a pH indicator. (pH 0.0-0.8/ pH 8.2-10.0).

Solubility
Soluble in ethanol, acetic acid and acetone. Insoluble in water.

Notes
Incompatible with strong oxidizing agents, acids, water reactive substances, azo, diazo, hydrazines, metals, nitrides, peroxides, hydroperoxides, epoxides and reducing agents.

Chemical Identifiers

CAS 145-50-6
Molecular Formula C27H18O2
Molecular Weight (g/mol) 374.439
MDL Number MFCD00078492
InChI Key VDDWRTZCUJCDJM-PNHLSOANSA-N
PubChem CID 5941340
IUPAC Name (4Z)-4-[(4-hydroxynaphthalen-1-yl)-phenylmethylidene]naphthalen-1-one
SMILES C1=CC=C(C=C1)C(=C2C=CC(=O)C3=CC=CC=C23)C4=CC=C(C5=CC=CC=C54)O

Specifications

Melting Point ∼225°C (decomposition)
Quantity 5 g
Beilstein 3471575
Synonym p-naphtholbenzein, 1-naphtholbenzein, alpha-naphtholbenzein, .alpha.-naphtholbenzein, 4-4-hydroxynaphthyl phenylmethylene naphthalen-1-one, naphtholbenzein, 1 4h-naphthalenone, 4-4-hydroxy-1-naphthalenyl phenylmethylene, 1 4h-naphthalenone,4-4-hydroxy-1-naphthalenyl phenylmethylene, 1 4h-naphthalenone, 4-.alpha.-4-hydroxy-1-naphthyl benzylidene
Solubility Information Soluble in ethanol,acetic acid and acetone. Insoluble in water.
Formula Weight 374.44
Chemical Name or Material alpha-Naphtholbenzein
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 205-656-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only