missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Methylene-gamma-valerolactone (stabilized with HQ) 97.0+%, TCI America™
$212.51 - $850.02
Chemical Identifiers
| CAS | 62873-16-9 |
|---|---|
| Molecular Formula | C6H8O2 |
| Molecular Weight (g/mol) | 112.13 |
| MDL Number | MFCD06797118 |
| InChI Key | KYLUHLJIAMFYKW-UHFFFAOYNA-N |
| Synonym | 4,5-Dihydro-5-methyl-3-methylene-2(3H)-furanone |
| PubChem CID | 99939 |
| IUPAC Name | 5-methyl-3-methylideneoxolan-2-one |
| SMILES | CC1CC(=C)C(=O)O1 |
Chemical Identifiers
| 62873-16-9 | |
| 112.13 | |
| KYLUHLJIAMFYKW-UHFFFAOYNA-N | |
| 99939 | |
| CC1CC(=C)C(=O)O1 |
| C6H8O2 | |
| MFCD06797118 | |
| 4,5-Dihydro-5-methyl-3-methylene-2(3H)-furanone | |
| 5-methyl-3-methylideneoxolan-2-one |
Specifications
| 62873-16-9 | |
| C6H8O2 | |
| 5 g | |
| KYLUHLJIAMFYKW-UHFFFAOYNA-N | |
| 5-methyl-3-methylideneoxolan-2-one | |
| 99939 | |
| ≥97.0% (GC) | |
| alpha-Methylene-gamma-valerolactone (stabilized with HQ) |
| Yellow | |
| MFCD06797118 | |
| 4,5-Dihydro-5-methyl-3-methylene-2(3H)-furanone | |
| CC1CC(=C)C(=O)O1 | |
| 112.13 | |
| 112.13 | |
| Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator