missing translation for 'onlineSavingsMsg'
Learn More

alpha-Methylene-gamma-valerolactone (stabilized with HQ) 97.0+%, TCI America™

Supplier:  TCI America M145325G

 View more versions of this product

Catalog No. M145325G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

alpha-Methylene-gamma-valerolactone (stabilized with HQ)
Yellow
MFCD06797118
4,5-Dihydro-5-methyl-3-methylene-2(3H)-furanone
CC1CC(=C)C(=O)O1
112.13
112.13
Liquid
62873-16-9
C6H8O2
25 g
KYLUHLJIAMFYKW-UHFFFAOYNA-N
5-methyl-3-methylideneoxolan-2-one
99939
≥97.0% (GC)

Chemical Identifiers

62873-16-9
112.13
KYLUHLJIAMFYKW-UHFFFAOYNA-N
99939
CC1CC(=C)C(=O)O1
C6H8O2
MFCD06797118
4,5-Dihydro-5-methyl-3-methylene-2(3H)-furanone
5-methyl-3-methylideneoxolan-2-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions