missing translation for 'onlineSavingsMsg'
Learn More

alpha-Methylene-gamma-butyrolactone (stabilized with 2,6-Di-tert-butyl-p-cresol) 95.0+%, TCI America™

Supplier:  TCI America M090725G

 View more versions of this product

Catalog No. M090725G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

alpha-Methylene-gamma-butyrolactone (stabilized with 2,6-Di-tert-butyl-p-cresol)
Yellow
C5H6O2
25 g
GSLDEZOOOSBFGP-UHFFFAOYSA-N
3-methylideneoxolan-2-one
68352
98.10
Liquid
547-65-9
90°C
MFCD00005407
2-methylenebutyrolactone, alpha-methylene-gamma-butyrolactone, 3-methylenedihydro-2 3h-furanone, tulipalin a, tulipane, 2 3h-furanone, dihydro-3-methylene, dihydro-3-methylene-2 3h-furanone, alpha-methylene butyrolactone, 3-methylenedihydrofuran-2 3h-one, alpha-methylene gamma-butyrolactone
C=C1CCOC1=O
98.101
CHEBI:104120
≥95.0% (GC)

Chemical Identifiers

547-65-9
98.101
GSLDEZOOOSBFGP-UHFFFAOYSA-N
68352
3-methylideneoxolan-2-one
C5H6O2
MFCD00005407
2-methylenebutyrolactone, alpha-methylene-gamma-butyrolactone, 3-methylenedihydro-2 3h-furanone, tulipalin a, tulipane, 2 3h-furanone, dihydro-3-methylene, dihydro-3-methylene-2 3h-furanone, alpha-methylene butyrolactone, 3-methylenedihydrofuran-2 3h-one, alpha-methylene gamma-butyrolactone
CHEBI:104120
C=C1CCOC1=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : LU3580400

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions