accessibility menu, dialog, popup

UserName

CH2OO

CAS RN 547-65-9

IUPAC Name: 3-methylideneoxolan-2-one
Synonyms: tulipalin A
Molecular Weight (g/mol): 98.1
Molecular Formula: C5H6O2
InChi Key: GSLDEZOOOSBFGP-UHFFFAOYSA-N
SMILES: C=C1CCOC1=O
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (5)
  • (2)
  • (1)
  • (2)