missing translation for 'onlineSavingsMsg'
Learn More

alpha-Methylene-gamma-butyrolactone (stabilized with 2,6-Di-tert-butyl-p-cresol) 95.0+%, TCI America™

Catalog No. M09075G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M09075G 5 g
M09071G 1 g
M090725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M09075G Supplier TCI America Supplier No. M09075G

Chemical Identifiers

CAS 547-65-9
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.101
MDL Number MFCD00005407
InChI Key GSLDEZOOOSBFGP-UHFFFAOYSA-N
Synonym 2-methylenebutyrolactone, alpha-methylene-gamma-butyrolactone, 3-methylenedihydro-2 3h-furanone, tulipalin a, tulipane, 2 3h-furanone, dihydro-3-methylene, dihydro-3-methylene-2 3h-furanone, alpha-methylene butyrolactone, 3-methylenedihydrofuran-2 3h-one, alpha-methylene gamma-butyrolactone
PubChem CID 68352
ChEBI CHEBI:104120
IUPAC Name 3-methylideneoxolan-2-one
SMILES C=C1CCOC1=O

Specifications

Color Yellow
Boiling Point 90°C
Quantity 5 g
Formula Weight 98.10
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material alpha-Methylene-gamma-butyrolactone (stabilized with 2,6-Di-tert-butyl-p-cresol)
RTECSNumber LU3580400
TSCA No
Recommended Storage Refrigerator