missing translation for 'onlineSavingsMsg'
Learn More

alpha-Methylene-gamma-butyrolactone, 95%, stabilized, Thermo Scientific Chemicals

Catalog No. AC414260050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC414260050 5 g Glass bottle
AC414260010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC414260050 Supplier Thermo Scientific Chemicals Supplier No. 414260050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 547-65-9
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.1
MDL Number MFCD00005407
InChI Key GSLDEZOOOSBFGP-UHFFFAOYSA-N
Synonym 2-methylenebutyrolactone, alpha-methylene-gamma-butyrolactone, 3-methylenedihydro-2 3h-furanone, tulipalin a, tulipane, 2 3h-furanone, dihydro-3-methylene, dihydro-3-methylene-2 3h-furanone, alpha-methylene butyrolactone, 3-methylenedihydrofuran-2 3h-one, alpha-methylene gamma-butyrolactone
PubChem CID 68352
ChEBI CHEBI:104120
IUPAC Name 3-methylideneoxolan-2-one
SMILES C=C1CCOC1=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow or Pink
Density 1.1200g/mL
Boiling Point 88°C to 92°C (12.0 mmHg)
Flash Point 37°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4710 to 1.475
Quantity 5 g
Fieser 11,80
Specific Gravity 1.12
Merck Index 15, 6134
Stabilizer 2, 6-di-tert.-butyl-p-cresol
Formula Weight 98.1
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material α-Methylene-γ-butyrolactone, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
EINECSNumber 208-931-6

RUO – Research Use Only