missing translation for 'onlineSavingsMsg'
Learn More

alpha-Methylcinnamaldehyde 95.0+%, TCI America™

Catalog No. p-7132251
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M058425G 25 g
M0584500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M058425G Supplier TCI America Supplier No. M058425G
Only null left

Chemical Identifiers

CAS 101-39-3
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.19
MDL Number MFCD00006976
InChI Key VLUMOWNVWOXZAU-CLFYSBASSA-N
Synonym alpha-methylcinnamaldehyde, a-methylcinnamaldehyde, 2-propenal, 2-methyl-3-phenyl, 2-methyl-3-phenylprop-2-enal, e-2-methyl-3-phenylpropenal, 2-methyl-3-phenylacrylaldehyde, e-2-methyl-3-phenylacrylaldehyde, 2-methyl-3-phenyl-2-propenal, 2-methyl-3-phenylacrolein, alpha-methyl-trans-cinnamaldehyde
PubChem CID 5372813
IUPAC Name (2Z)-2-methyl-3-phenylprop-2-enal
SMILES C\C(C=O)=C\C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 132°C
Quantity 25 g
Formula Weight 146.19
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material alpha-Methylcinnamaldehyde
EINECSNumber (3)-4622
RTECSNumber GD6600000
TSCA Yes