accessibility menu, dialog, popup

UserName

CH3O(Z)

CAS RN 101-39-3

IUPAC Name: (2Z)-2-methyl-3-phenylprop-2-enal
Synonyms: methylcinnamic aldehyde
Molecular Weight (g/mol): 146.19
Molecular Formula: C10H10O
InChi Key: VLUMOWNVWOXZAU-CLFYSBASSA-N
SMILES: C\C(C=O)=C\C1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)