missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals alpha-Mating Factor (1-6)

Catalog No. AAJ66484MA
Change view
Click to view available options
Quantity:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66484MA 10 mg
AAJ66484MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ66484MA Supplier Thermo Scientific Chemicals Supplier No. J66484MA

CAS: 65418-88-4 | C45H59N11O8 | 882.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 65418-88-4

Specifications

Content And Storage Store at -20°C
Molecular Formula C45H59N11O8
Format Solid
InChI Key PBLMMVDDGREFKH-UHFFFAOYNA-N
SMILES CC(C)CC(NC(=O)C(CCC(N)=O)NC(=O)C(CC(C)C)NC(=O)C(CC1=CNC2=CC=CC=C12)NC(=O)C(CC1=CN=CN1)NC(=O)C(N)CC1=CNC2=CC=CC=C12)C(O)=O
IUPAC Name 2-{2-[2-(2-{2-[2-amino-3-(1H-indol-3-yl)propanamido]-3-(1H-imidazol-5-yl)propanamido}-3-(1H-indol-3-yl)propanamido)-4-methylpentanamido]-4-carbamoylbutanamido}-4-methylpentanoic acid
Molecular Weight (g/mol) 882.04
Quantity 10 mg

RUO – Research Use Only