accessibility menu, dialog, popup

UserName

H3CCH3ONHOH2NOHNH3CCH3OOHHNOHNNHONNHHNONHNH2

CAS RN 65418-88-4

IUPAC Name: 2-{2-[2-(2-{2-[2-amino-3-(1H-indol-3-yl)propanamido]-3-(1H-imidazol-5-yl)propanamido}-3-(1H-indol-3-yl)propanamido)-4-methylpentanamido]-4-carbamoylbutanamido}-4-methylpentanoic acid
Synonyms: 2-{2-[2-(2-{2-[2-amino-3-(1H-indol-3-yl)propanamido]-3-(3H-imidazol-4-yl)propanamido}-3-(1H-indol-3-yl)propanamido)-4-methylpentanamido]-4-carbamoylbutanamido}-4-methylpentanoic acid
Molecular Weight (g/mol): 882.04
Molecular Formula: C45H59N11O8
InChi Key: PBLMMVDDGREFKH-UHFFFAOYNA-N
SMILES: CC(C)CC(NC(=O)C(CCC(N)=O)NC(=O)C(CC(C)C)NC(=O)C(CC1=CNC2=CC=CC=C12)NC(=O)C(CC1=CN=CN1)NC(=O)C(N)CC1=CNC2=CC=CC=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)