missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-iso-Methylionone 70.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 25 mL |
| Formula Weight | 206.33 |
| Percent Purity | ≥70.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | alpha-iso-Methylionone |
Safety and Handling
| EINECSNumber | (3)-2385 |
| TSCA | Yes |