Learn More
alpha-Fluorophenylacetic acid, 97%
CAS: 1578-63-8 | C8H7FO2 | 154.14 g/mol
Supplier: Thermo Scientific Chemicals L01745ME
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| alpha-Fluorophenylacetic acid | |
| 78°C to 82°C | |
| MFCD00004221 | |
| UN2811 | |
| alpha-fluorophenylacetic acid, fluorophenylacetic acid, fluoro phenyl acetic acid, .alpha.-fluorophenylacetic acid, benzeneacetic acid, .alpha.-fluoro, fluorophenyl acetic acid, a-fluorophenylacetic acid, a-fluorobenzeneacetic acid, a-fluorophenyl acetic acid, acmc-1bt6i | |
| C1=CC=C(C=C1)C(C(=O)O)F | |
| 154.14 | |
| 154.14 |
| 1578-63-8 | |
| C8H7FO2 | |
| 500 mg | |
| 2087378 | |
| ATPPNMLQNZHDOG-UHFFFAOYSA-N | |
| 2-fluoro-2-phenylacetic acid | |
| 102649 | |
| 98% |
Chemical Identifiers
| 1578-63-8 | |
| 154.14 | |
| ATPPNMLQNZHDOG-UHFFFAOYSA-N | |
| 102649 | |
| C1=CC=C(C=C1)C(C(=O)O)F |
| C8H7FO2 | |
| MFCD00004221 | |
| alpha-fluorophenylacetic acid, fluorophenylacetic acid, fluoro phenyl acetic acid, .alpha.-fluorophenylacetic acid, benzeneacetic acid, .alpha.-fluoro, fluorophenyl acetic acid, a-fluorophenylacetic acid, a-fluorobenzeneacetic acid, a-fluorophenyl acetic acid, acmc-1bt6i | |
| 2-fluoro-2-phenylacetic acid |
Safety and Handling
GHS H Statement
H301-H311-H331-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P361-P363-P501c
H301+H311-H315-H319-H330-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 216-420-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only