missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha,epsilon-Dibenzoyl-DL-lysine 99.0+%, TCI America™
$157.70 - $382.51
Chemical Identifiers
| CAS | 32513-92-1 |
|---|---|
| Molecular Formula | C20H22N2O4 |
| MDL Number | MFCD00059684 |
Chemical Identifiers
| 32513-92-1 | |
| C20H22N2O4 | |
| MFCD00059684 |
Specifications
| 32513-92-1 | |
| White-Yellow | |
| MFCD00059684 | |
| 354.41 | |
| Crystalline Powder |
| 146°C | |
| C20H22N2O4 | |
| 1 g | |
| ≥99.0% (T) | |
| alpha,epsilon-Dibenzoyl-DL-lysine |
Safety and Handling
TSCA : No