missing translation for 'onlineSavingsMsg'
Learn More

alpha,epsilon-Dibenzoyl-DL-lysine 99.0+%, TCI America™

Catalog No. b02085g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B02085G 5 g
B02081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B02085G Supplier TCI America Supplier No. B02085G

Chemical Identifiers

CAS 32513-92-1
Molecular Formula C20H22N2O4
MDL Number MFCD00059684

Specifications

Melting Point 146°C
Color White-Yellow
Quantity 5 g
Formula Weight 354.41
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material alpha,epsilon-Dibenzoyl-DL-lysine