missing translation for 'onlineSavingsMsg'
Learn More

alpha-D-Ribose 1,5-Bis(phosphate) Tetrasodium Salt, TCI America™

Catalog No. r00825mg
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R00825MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R00825MG Supplier TCI America Supplier No. R00825MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 113599-17-0
Molecular Formula C5H8Na4O11P2
Molecular Weight (g/mol) 398.015
InChI Key NOGFWTMOJJPOTG-HPORTLBWSA-J
PubChem CID 14035694
IUPAC Name tetrasodium;[(2R,3R,4S,5R)-3,4-dihydroxy-5-(phosphonatooxymethyl)oxolan-2-yl] phosphate
SMILES C(C1C(C(C(O1)OP(=O)([O-])[O-])O)O)OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+]

Specifications

Quantity 5 mg
Formula Weight 398.01
Chemical Name or Material alpha-D-Ribose 1,5-Bis(phosphate) Tetrasodium Salt
TSCA No
Recommended Storage Freezer