missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ alpha-D-Glucose pentaacetate, 98%

Chemical Identifiers

CAS 604-68-2
Molecular Formula C16H22O11
Molecular Weight (g/mol) 390.34
MDL Number MFCD00064071,MFCD00064081
InChI Key LPTITAGPBXDDGR-UHFFFAOYNA-N
Synonym alpha-d-glucose pentaacetate, a-d-glucopyranose pentaacetate, pentaacetyl-alpha-d-glucose, penta-o-acetyl-alpha-d-glucopyranose, unii-1cs424ns93, 1,2,3,4,6-penta-o-acetyl-alpha-d-glucopyranose, alpha-d-glucopyranose pentaacetate, 2r,3r,4s,5r,6r-3,4,5,6-tetrakis acetyloxy oxan-2-yl methyl acetate, glucopyranose, pentaacetate, .alpha.-d, a-d-glucose pentaacetate
PubChem CID 2723636
IUPAC Name [(2R,3R,4S,5R,6R)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC119910250
SDS View Documents
Thermo Scientific™
119910250
25g Plastic bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC119911000
SDS View Documents
Thermo Scientific™
119911000
100g Plastic bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

604-68-2
390.34
LPTITAGPBXDDGR-UHFFFAOYNA-N
2723636
CC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O
C16H22O11
MFCD00064071,MFCD00064081
alpha-d-glucose pentaacetate, a-d-glucopyranose pentaacetate, pentaacetyl-alpha-d-glucose, penta-o-acetyl-alpha-d-glucopyranose, unii-1cs424ns93, 1,2,3,4,6-penta-o-acetyl-alpha-d-glucopyranose, alpha-d-glucopyranose pentaacetate, 2r,3r,4s,5r,6r-3,4,5,6-tetrakis acetyloxy oxan-2-yl methyl acetate, glucopyranose, pentaacetate, .alpha.-d, a-d-glucose pentaacetate
[(2R,3R,4S,5R,6R)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate

Specifications

604-68-2
100.0
Authentic
Plastic bottle
MFCD00064071,MFCD00064081
31, 119
+ 97.00
Solubility in water: < 5g/L (25°C). Other solubilities: slightly soluble in ethanol,, soluble in diethyl ether, chloroform, acetic acid
CC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O
390.34
390.33
α-D-Glucose pentaacetate, 98%
97.5
>150°C
97.5% min. (GC)
C16H22O11
25g
+97° (20°C c=1,CHC3)
alpha-d-glucose pentaacetate, a-d-glucopyranose pentaacetate, pentaacetyl-alpha-d-glucose, penta-o-acetyl-alpha-d-glucopyranose, unii-1cs424ns93, 1,2,3,4,6-penta-o-acetyl-alpha-d-glucopyranose, alpha-d-glucopyranose pentaacetate, 2r,3r,4s,5r,6r-3,4,5,6-tetrakis acetyloxy oxan-2-yl methyl acetate, glucopyranose, pentaacetate, .alpha.-d, a-d-glucose pentaacetate
LPTITAGPBXDDGR-UHFFFAOYNA-N
[(2R,3R,4S,5R,6R)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate
2723636
98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 210-073-2

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only