missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals alpha-D-Glucose pentaacetate, 98%

Catalog No. ac119911000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC119911000 100 g Plastic bottle
AC119910250 25 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC119911000 Supplier Thermo Scientific Chemicals Supplier No. 119911000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 604-68-2 | C16H22O11 | 390.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 604-68-2
Molecular Formula C16H22O11
Molecular Weight (g/mol) 390.34
MDL Number MFCD00064071,MFCD00064081
InChI Key LPTITAGPBXDDGR-UHFFFAOYNA-N
Synonym alpha-d-glucose pentaacetate, a-d-glucopyranose pentaacetate, pentaacetyl-alpha-d-glucose, penta-o-acetyl-alpha-d-glucopyranose, unii-1cs424ns93, 1,2,3,4,6-penta-o-acetyl-alpha-d-glucopyranose, alpha-d-glucopyranose pentaacetate, 2r,3r,4s,5r,6r-3,4,5,6-tetrakis acetyloxy oxan-2-yl methyl acetate, glucopyranose, pentaacetate, .alpha.-d, a-d-glucose pentaacetate
PubChem CID 2723636
IUPAC Name [(2R,3R,4S,5R,6R)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 109.0°C to 113.0°C
Color White
Flash Point >150°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Quantity 100 g
Beilstein 31, 119
Specific Rotation Condition +97° (20°C c=1,CHC3)
Specific Rotation + 97.00
Solubility Information Solubility in water: < 5g/L (25°C). Other solubilities: slightly soluble in ethanol,, soluble in diethyl ether, chloroform, acetic acid
Formula Weight 390.33
Percent Purity 98%
Physical Form Powder
Chemical Name or Material α-D-Glucose pentaacetate, 98%
Show More Show Less
EINECSNumber 210-073-2
TSCA TSCA

RUO – Research Use Only