missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals alpha-D(+)-Glucose, 99+%, anhydrous

Catalog No. AC170080250
Change view
Click to view available options
Quantity:
2.5 kg
25 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC170080250 25 kg
AC170080025 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC170080250 Supplier Thermo Scientific Chemicals Supplier No. 170080250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 492-62-6 | C6H12O6 | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 492-62-6
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
MDL Number MFCD00063774
InChI Key WQZGKKKJIJFFOK-DVKNGEFBSA-N
Synonym alpha-d-glucose, alpha-d-glucopyranose, alpha-dextrose, alpha-glucose, glucopyranose, alpha-d, alpha-d-glc, unii-5j5i9eb41e, glucoses, grape sugar, d-gluco-hexose
PubChem CID 79025
ChEBI CHEBI:17925
IUPAC Name (2S,3R,4S,5S,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol
SMILES C(C1C(C(C(C(O1)O)O)O)O)O

Specifications

Acidity 0.15mL 0.1N NaOH/6g max.
CAS Min % 99.0
CAS Max % 100.0
Melting Point 156.0°C to 158.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (ELSD) (HPLC)
Quantity 25 kg
Beilstein 31, 83
Merck Index 14, 4459
Specific Rotation Condition + 52.90 (20.00°C c=10,water,ammonia)
Specific Rotation + 52.90
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in alcohol
Formula Weight 180.16
Percent Purity 99+%
Related Substances 0.15% max. (Fructose)
Physical Form Fine Crystalline Powder
Chemical Name or Material α-D(+)-Glucose, Anhydrous
Show More Show Less
EINECSNumber 207-757-8

RUO – Research Use Only