missing translation for 'onlineSavingsMsg'
Learn More

alpha-Cyclopentyl-DL-mandelic Acid 98.0+%, TCI America™

Catalog No. C23945G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23945G 5 g
C239425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23945G Supplier TCI America Supplier No. C23945G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 427-49-6
Molecular Formula C13H16O3
Molecular Weight (g/mol) 220.268
MDL Number MFCD00019296
InChI Key WFLUEQCOAQCQLP-UHFFFAOYSA-N
Synonym alpha-cyclopentylmandelic acid, cyclopentylphenylglycolic acid, cyclopentyl hydroxy phenylacetic acid, cyclopentyl-hydroxy-phenyl-acetic acid, cyclopentyl mandelic acid, alpha-cyclopentyl-dl-mandelic acid, a-cyclo pentyl mandelic acid, alpha-cyclopentylmandelicacid, 2-rs-cyclopentyl-2-hydroxy-2-phenylacetic acid alpha-cyclopentylmandelic acid, cyclopentylmandelic acid
PubChem CID 98283
IUPAC Name 2-cyclopentyl-2-hydroxy-2-phenylacetic acid
SMILES C1CCC(C1)C(C2=CC=CC=C2)(C(=O)O)O

Specifications

Melting Point 148°C
Color White-Yellow
Quantity 5 g
Formula Weight 220.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material alpha-Cyclopentyl-DL-mandelic Acid