accessibility menu, dialog, popup

UserName

CAS RN 427-49-6

IUPAC Name: 2-cyclopentyl-2-hydroxy-2-phenylacetic acid
Synonyms: cyclopentyl(hydroxy)phenylacetic acid
Molecular Weight (g/mol): 220.27
Molecular Formula: C13H16O3
InChi Key: WFLUEQCOAQCQLP-UHFFFAOYNA-N
SMILES: OC(=O)C(O)(C1CCCC1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)