missing translation for 'onlineSavingsMsg'
Learn More

alpha-Cyanocinnamic Acid 98.0+%, TCI America™

Catalog No. C044725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C044725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C044725G Supplier TCI America Supplier No. C044725G
Only null left

Chemical Identifiers

CAS 1011-92-3
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.171
MDL Number MFCD00016813
InChI Key CDUQMGQIHYISOP-UHFFFAOYSA-N
Synonym 2-cyano-3-phenylacrylic acid, alpha-cyanocinnamicacid, cyanocinnamic acid, 2-cyano-3-phenyl-2-propenoic acid, acmc-1b4v0, a-cyano-b-phenylacrylic acid, alpha-cyano-beta-phenyl-acrylic acid
PubChem CID 70534
IUPAC Name 2-cyano-3-phenylprop-2-enoic acid
SMILES C1=CC=C(C=C1)C=C(C#N)C(=O)O

Specifications

Melting Point 181°C
Color White-Yellow
Quantity 25 g
Formula Weight 173.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material alpha-Cyanocinnamic Acid
RTECSNumber GD8562500
TSCA No