accessibility menu, dialog, popup

UserName

CAS RN 1011-92-3

IUPAC Name: (2Z)-2-cyano-3-phenylprop-2-enoic acid
Synonyms: (2Z)-2-cyano-3-phenylprop-2-enoic acid
Molecular Weight (g/mol): 173.17
Molecular Formula: C10H7NO2
InChi Key: CDUQMGQIHYISOP-TWGQIWQCSA-N
SMILES: OC(=O)C(=C/C1=CC=CC=C1)\C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results