missing translation for 'onlineSavingsMsg'
Learn More

alpha-Cyano-o-tolunitrile, 80%, Thermo Scientific™

Catalog No. p-94889
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC152570250 25 g Glass bottle
AC152571000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC152570250 Supplier Thermo Scientific Chemicals Supplier No. 152570250
Only null left

Chemical Identifiers

CAS 3759-28-2
Molecular Formula C9H6N2
Molecular Weight (g/mol) 142.16
MDL Number MFCD00001895
InChI Key GKHSEDFDYXZGCG-UHFFFAOYSA-N
Synonym 2-cyanomethyl benzonitrile, homophthalonitrile, 2-cyanobenzyl cyanide, 2-cyanophenylacetonitrile, alpha-cyano-o-tolunitrile, o-cyanobenzyl cyanide, benzeneacetonitrile, 2-cyano, 2-cyanobenzeneacetonitrile, o-cyanophenyl acetonitrile, o-tolunitrile, alpha-cyano
PubChem CID 77368
IUPAC Name 2-(cyanomethyl)benzonitrile
SMILES N#CCC1=CC=CC=C1C#N

Specifications

CAS Min % 75.0
CAS Max % 100.0
Melting Point 76°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 75% min. (GC)
Packaging Glass bottle
Linear Formula NCCH2C6H4CN
Quantity 25 g
Beilstein 09, 859
Formula Weight 142.16
Percent Purity 80%
Grade Technical
Physical Form Chunks and/or Powder
Chemical Name or Material α-Cyano-o-tolunitrile, tech.
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Fatal if inhaled.
Toxic in contact with skin.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 223-167-3
RTECSNumber XV0930000
TSCA TSCA

RUO – Research Use Only