missing translation for 'onlineSavingsMsg'
Learn More

alpha-Chloro-p-xylene 98.0+%, TCI America™

Catalog No. C031225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C031225G 25 g
C0312500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C031225G Supplier TCI America Supplier No. C031225G
Only null left

Chemical Identifiers

CAS 104-82-5
Molecular Formula C8H9Cl
Molecular Weight (g/mol) 140.61
MDL Number MFCD00000919
InChI Key DMHZDOTYAVHSEH-UHFFFAOYSA-N
Synonym 4-methylbenzyl chloride, p-xylyl chloride, 1-chloromethyl-4-methylbenzene, alpha-chloro-p-xylene, p-methylbenzyl chloride, benzene, 1-chloromethyl-4-methyl, p-chloromethyl toluene, 4-chloromethyl toluene, p-tolylmethyl chloride, p-xylene, alpha-chloro
PubChem CID 7722
IUPAC Name 1-(chloromethyl)-4-methylbenzene
SMILES CC1=CC=C(C=C1)CCl

Specifications

Color Colorless
Boiling Point 202°C
Quantity 25 g
UN Number 3265
Formula Weight 140.61
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material alpha-Chloro-p-xylene
EINECSNumber (3)-2691
RTECSNumber ZE4030000
TSCA Yes