missing translation for 'onlineSavingsMsg'
Learn More

alpha-Chloro-p-xylene, 98%

Catalog No. p-92020
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC110171000 100 mL Glass bottle
AC110175000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC110171000 Supplier Thermo Scientific Chemicals Supplier No. 110171000
Only null left

CAS: 104-82-5 | C8H9Cl | 140.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104-82-5
Molecular Formula C8H9Cl
Molecular Weight (g/mol) 140.61
MDL Number MFCD00000919
InChI Key DMHZDOTYAVHSEH-UHFFFAOYSA-N
Synonym 4-methylbenzyl chloride, p-xylyl chloride, 1-chloromethyl-4-methylbenzene, alpha-chloro-p-xylene, p-methylbenzyl chloride, benzene, 1-chloromethyl-4-methyl, p-chloromethyl toluene, 4-chloromethyl toluene, p-tolylmethyl chloride, p-xylene, alpha-chloro
PubChem CID 7722
IUPAC Name 1-(chloromethyl)-4-methylbenzene
SMILES CC1=CC=C(C=C1)CCl

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 4°C
Color Colorless to Yellow
Density 1.0600g/mL
Boiling Point 200°C to 202°C
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 97.5 % min. (GC)
Packaging Glass bottle
Refractive Index 1.5330 to 1.535
Linear Formula CH3C6H4CH2Cl
Quantity 100 mL
Beilstein 05, 384
Specific Gravity 1.06
Merck Index 15, 10289
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with absolute alcohol and ether
Formula Weight 140.61
Percent Purity 98%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material α-Chloro-p-xylene, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water
EINECSNumber 203-241-1
RTECSNumber ZE4030000
TSCA TSCA

RUO – Research Use Only