missing translation for 'onlineSavingsMsg'
Learn More

alpha-Chloro-o-xylene, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 110151000

 View more versions of this product

Catalog No. AC110151000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

α-Chloro-o-xylene
98.5
1.0800g/mL
73°C
98.5% min. (GC)
C8H9Cl
CH3C6H4CH2Cl
100mL
1.08
2-methylbenzyl chloride, 1-chloromethyl-2-methylbenzene, o-xylyl chloride, alpha-chloro-o-xylene, o-methylbenzyl chloride, o-chloromethyl toluene, benzene, 1-chloromethyl-2-methyl, 2-methylbenzylchloride, 2-chloromethyl toluene, unii-eqt3v044tw
VQRBXYBBGHOGFT-UHFFFAOYSA-N
1-(chloromethyl)-2-methylbenzene
11093
99%
552-45-4
100.0
197.0°C to 199.0°C
Authentic
Glass bottle
1.5405 to 1.5425
MFCD00000899
05,364
15,10289
Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,ketones and,chlorinated hydrocarbons
CC1=CC=CC=C1CCl
140.61
140.61

Chemical Identifiers

552-45-4
140.61
VQRBXYBBGHOGFT-UHFFFAOYSA-N
11093
CC1=CC=CC=C1CCl
C8H9Cl
MFCD00000899
2-methylbenzyl chloride, 1-chloromethyl-2-methylbenzene, o-xylyl chloride, alpha-chloro-o-xylene, o-methylbenzyl chloride, o-chloromethyl toluene, benzene, 1-chloromethyl-2-methyl, 2-methylbenzylchloride, 2-chloromethyl toluene, unii-eqt3v044tw
1-(chloromethyl)-2-methylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.

GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously

GHS Signal Word: Danger

EINECSNumber : 209-013-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only