missing translation for 'onlineSavingsMsg'
Learn More

alpha-Chloro-m-xylene 97.0+%, TCI America™

Catalog No. p-7124653
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C086525G 25 g
C0865500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C086525G Supplier TCI America Supplier No. C086525G
Only null left

Chemical Identifiers

CAS 620-19-9
Molecular Formula C8H9Cl
Molecular Weight (g/mol) 140.61
MDL Number MFCD00000909
InChI Key LZBOHNCMCCSTJX-UHFFFAOYSA-N
Synonym 3-methylbenzyl chloride, 1-chloromethyl-3-methylbenzene, m-xylyl chloride, 3-methylbenzylchloride, alpha-chloro-m-xylene, m-methylbenzyl chloride, 1-chloromethyl-3-methyl-benzene, m-chloromethyl toluene, 3-chloromethyl toluene, benzene, 1-chloromethyl-3-methyl
PubChem CID 12102
IUPAC Name 1-(chloromethyl)-3-methylbenzene
SMILES CC1=CC(=CC=C1)CCl

Specifications

Color Yellow
Boiling Point 196°C
Quantity 25 g
UN Number 3265
Formula Weight 140.61
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material alpha-Chloro-m-xylene
EINECSNumber (3)-2691
TSCA No