missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Bromo-m-xylene 98.0+%, TCI America™
$380.27 - $1895.75
Chemical Identifiers
| CAS | 620-13-3 |
|---|---|
| Molecular Formula | C8H9Br |
| Molecular Weight (g/mol) | 185.064 |
| MDL Number | MFCD00000177 |
| InChI Key | FWLWTILKTABGKQ-UHFFFAOYSA-N |
| Synonym | 3-methylbenzyl bromide, 1-bromomethyl-3-methylbenzene, alpha-bromo-m-xylene, m-methylbenzyl bromide, m-xylyl bromide, benzene, 1-bromomethyl-3-methyl, 3-bromomethyl toluene, m-xylene, .alpha.-bromo, 3-methylbenzylbromide, m-xylene, alpha-bromo |
| PubChem CID | 12099 |
| IUPAC Name | 1-(bromomethyl)-3-methylbenzene |
| SMILES | CC1=CC(=CC=C1)CBr |
Chemical Identifiers
| 620-13-3 | |
| 185.064 | |
| FWLWTILKTABGKQ-UHFFFAOYSA-N | |
| 12099 | |
| CC1=CC(=CC=C1)CBr |
| C8H9Br | |
| MFCD00000177 | |
| 3-methylbenzyl bromide, 1-bromomethyl-3-methylbenzene, alpha-bromo-m-xylene, m-methylbenzyl bromide, m-xylyl bromide, benzene, 1-bromomethyl-3-methyl, 3-bromomethyl toluene, m-xylene, .alpha.-bromo, 3-methylbenzylbromide, m-xylene, alpha-bromo | |
| 1-(bromomethyl)-3-methylbenzene |
Specifications
| 620-13-3 | |
| 98°C | |
| MFCD00000177 | |
| 1701 | |
| FWLWTILKTABGKQ-UHFFFAOYSA-N | |
| 1-(bromomethyl)-3-methylbenzene | |
| 12099 | |
| ≥98.0% (GC) | |
| alpha-Bromo-m-xylene |
| Yellow | |
| C8H9Br | |
| 5 g | |
| 3-methylbenzyl bromide, 1-bromomethyl-3-methylbenzene, alpha-bromo-m-xylene, m-methylbenzyl bromide, m-xylyl bromide, benzene, 1-bromomethyl-3-methyl, 3-bromomethyl toluene, m-xylene, .alpha.-bromo, 3-methylbenzylbromide, m-xylene, alpha-bromo | |
| CC1=CC(=CC=C1)CBr | |
| 185.064 | |
| 185.06 | |
| Liquid |
Safety and Handling
RTECSNumber : CY9039200
TSCA : Yes