missing translation for 'onlineSavingsMsg'
Learn More

alpha-Bromo-4-fluorophenylacetic acid, 96%

Catalog No. p-7057234
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6669203 1 g
AAH6669206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6669203 Supplier Thermo Scientific Chemicals Supplier No. H6669203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 29270-33-5 | C8H6BrFO2 | 233.036 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 29270-33-5
Molecular Formula C8H6BrFO2
Molecular Weight (g/mol) 233.036
MDL Number MFCD08276759
InChI Key ATILMAUMZRFROS-UHFFFAOYSA-N
Synonym 2-bromo-2-4-fluorophenyl acetic acid, alpha-bromo-4-fluorophenylacetic acid, bromo 4-fluorophenyl acetic acid, alpha-bromo-4-fluorophenyl acetic acid, 2-bromanyl-2-4-fluorophenyl ethanoic acid, alpha-bromo-4-fluorophenylacetic acid 9
PubChem CID 16217752
IUPAC Name 2-bromo-2-(4-fluorophenyl)acetic acid
SMILES C1=CC(=CC=C1C(C(=O)O)Br)F

Specifications

Melting Point 96°C to 100°C
Color White to Brown
Quantity 1 g
UN Number UN3261
Formula Weight 233.03
Percent Purity 96%
Physical Form Powder
Chemical Name or Material alpha-Bromo-4-fluorophenylacetic acid
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only