Learn More
alpha-Bromo-4-chlorophenylacetic acid, 97%
CAS: 3381-73-5 | C8H6BrClO2 | 249.49 g/mol
$77.32 - $261.26
Chemical Identifiers
| CAS | 3381-73-5 |
|---|---|
| Molecular Formula | C8H6BrClO2 |
| Molecular Weight (g/mol) | 249.49 |
| MDL Number | MFCD08276760 |
| InChI Key | KKOAAWLOOHBFQP-UHFFFAOYNA-N |
| Synonym | 2-bromo-2-4-chlorophenyl acetic acid, alpha-bromo-4-chlorophenylacetic acid, bromo 4-chlorophenyl acetic acid, acmc-1ct0u, bromo-4-chlorophenyl acetic acid, bromo-4-chloro-phenyl acetic acid, bromo-4-chloro-phenyl-acetic acid, 2-bromo-2-4-chlorophenyl aceticacid, alpha-bromo-p-chloro phenylacetic acid, alpha-bromo-p-chlorophenyl acetic acid |
| PubChem CID | 10490868 |
| IUPAC Name | 2-bromo-2-(4-chlorophenyl)acetic acid |
| SMILES | OC(=O)C(Br)C1=CC=C(Cl)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6014103
|
Thermo Scientific Chemicals
H6014103 |
1 g |
Each for $77.32
|
|
|||||
|
AAH6014106
|
Thermo Scientific Chemicals
H6014106 |
5 g |
Each for $261.26
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 3381-73-5 | |
| 249.49 | |
| KKOAAWLOOHBFQP-UHFFFAOYNA-N | |
| 10490868 | |
| OC(=O)C(Br)C1=CC=C(Cl)C=C1 |
| C8H6BrClO2 | |
| MFCD08276760 | |
| 2-bromo-2-4-chlorophenyl acetic acid, alpha-bromo-4-chlorophenylacetic acid, bromo 4-chlorophenyl acetic acid, acmc-1ct0u, bromo-4-chlorophenyl acetic acid, bromo-4-chloro-phenyl acetic acid, bromo-4-chloro-phenyl-acetic acid, 2-bromo-2-4-chlorophenyl aceticacid, alpha-bromo-p-chloro phenylacetic acid, alpha-bromo-p-chlorophenyl acetic acid | |
| 2-bromo-2-(4-chlorophenyl)acetic acid |
Specifications
| 95°C to 99°C | |
| 1 g | |
| MFCD08276760 | |
| 2-bromo-2-4-chlorophenyl acetic acid, alpha-bromo-4-chlorophenylacetic acid, bromo 4-chlorophenyl acetic acid, acmc-1ct0u, bromo-4-chlorophenyl acetic acid, bromo-4-chloro-phenyl acetic acid, bromo-4-chloro-phenyl-acetic acid, 2-bromo-2-4-chlorophenyl aceticacid, alpha-bromo-p-chloro phenylacetic acid, alpha-bromo-p-chlorophenyl acetic acid | |
| OC(=O)C(Br)C1=CC=C(Cl)C=C1 | |
| 249.49 | |
| 249.49 | |
| alpha-Bromo-4-chlorophenylacetic acid |
| 3381-73-5 | |
| C8H6BrClO2 | |
| UN3261 | |
| KKOAAWLOOHBFQP-UHFFFAOYNA-N | |
| 2-bromo-2-(4-chlorophenyl)acetic acid | |
| 10490868 | |
| 97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only