missing translation for 'onlineSavingsMsg'
Learn More

alpha-Bisabolol, 96%

CAS: 515-69-5 | C15H26O | 222.372 g/mol

$115.99 - $416.62

Chemical Identifiers

CAS 515-69-5
Molecular Formula C15H26O
Molecular Weight (g/mol) 222.372
MDL Number MFCD03846910
InChI Key RGZSQWQPBWRIAQ-LSDHHAIUSA-N
Synonym bisabolol, alpha-bisabolol, +-alpha-bisabolol, d-alpha-bisabolol, .alpha.-bisabolol, +-1'r,2r-alpha-bisabolol, 2r-6-methyl-2-1r-4-methylcyclohex-3-en-1-yl hept-5-en-2-ol, 3-cyclohexene-1-methanol, .alpha.,4-dimethyl-.alpha.-4-methyl-3-pentenyl-, r*,r*, dsstox_cid_25964, dsstox_rid_81260
PubChem CID 1549992
IUPAC Name (2R)-6-methyl-2-[(1R)-4-methylcyclohex-3-en-1-yl]hept-5-en-2-ol
SMILES CC1=CCC(CC1)C(C)(CCC=C(C)C)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2145709
View Documents
Thermo Scientific Chemicals
B2145709
10 g
Each for $115.99
Only null left
 
AAB2145718
View Documents
Thermo Scientific Chemicals
B2145718
50 g
Each for $416.62
Only null left
 
Description

Description

It finds its application as a masking agent, skin conditioning agent and soothing agent in cosmetics industry. The most important effects of Bisabolol for the use in cosmetics are anti-inflammatory, wound-healing, anti-bacterial and anti-mycotic. Bisabolol is therefore perfectly suited for the use in all kinds of skin-care products It can be used as active substance in cosmetic preparations for the protection and care of sensitive skin, preparations for babies and children, sunscreen and after-sun products, aftershaves and preparations for the oral hygiene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as a masking agent, skin conditioning agent and soothing agent in cosmetics industry. The most important effects of Bisabolol for the use in cosmetics are anti-inflammatory, wound-healing, anti-bacterial and anti-mycotic. Bisabolol is therefore perfectly suited for the use in all kinds of skin-care products It can be used as active substance in cosmetic preparations for the protection and care of sensitive skin, preparations for babies and children, sunscreen and after-sun products, aftershaves and preparations for the oral hygiene.

Solubility
Soluble in alcohol, natural, mineral and synthetic oils. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

515-69-5
222.372
RGZSQWQPBWRIAQ-LSDHHAIUSA-N
1549992
CC1=CCC(CC1)C(C)(CCC=C(C)C)O
C15H26O
MFCD03846910
bisabolol, alpha-bisabolol, +-alpha-bisabolol, d-alpha-bisabolol, .alpha.-bisabolol, +-1'r,2r-alpha-bisabolol, 2r-6-methyl-2-1r-4-methylcyclohex-3-en-1-yl hept-5-en-2-ol, 3-cyclohexene-1-methanol, .alpha.,4-dimethyl-.alpha.-4-methyl-3-pentenyl-, r*,r*, dsstox_cid_25964, dsstox_rid_81260
(2R)-6-methyl-2-[(1R)-4-methylcyclohex-3-en-1-yl]hept-5-en-2-ol

Specifications

515-69-5
135°C (275°F)
1.495
10 g
Air Sensitive
bisabolol, alpha-bisabolol, +-alpha-bisabolol, d-alpha-bisabolol, .alpha.-bisabolol, +-1'r,2r-alpha-bisabolol, 2r-6-methyl-2-1r-4-methylcyclohex-3-en-1-yl hept-5-en-2-ol, 3-cyclohexene-1-methanol, .alpha.,4-dimethyl-.alpha.-4-methyl-3-pentenyl-, r*,r*, dsstox_cid_25964, dsstox_rid_81260
RGZSQWQPBWRIAQ-LSDHHAIUSA-N
(2R)-6-methyl-2-[(1R)-4-methylcyclohex-3-en-1-yl]hept-5-en-2-ol
1549992
96%
Bisabolol
0.93
C15H26O
MFCD03846910
5733954
14,1241
Soluble in alcohol,natural,mineral and synthetic oils. Insoluble in water.
CC1=CCC(CC1)C(C)(CCC=C(C)C)O
222.372
222.37
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 208-205-9

RTECSNumber : MJ9685000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only