missing translation for 'onlineSavingsMsg'
Learn More

alpha-Asarone 98.0+%, TCI America™

Catalog No. A25905G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25905G 5 g
A25901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A25905G Supplier TCI America Supplier No. A25905G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2883-98-9
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.257
MDL Number MFCD00064457
InChI Key RKFAZBXYICVSKP-AATRIKPKSA-N
Synonym alpha-asarone, asarone, trans-isoasarone, trans-asarone, asaron, azaron, etherophenol, trans-isoasaron, e-asarone, asarum camphor
PubChem CID 636822
ChEBI CHEBI:78309
IUPAC Name 1,2,4-trimethoxy-5-[(E)-prop-1-enyl]benzene
SMILES CC=CC1=CC(=C(C=C1OC)OC)OC

Specifications

Melting Point 61°C
Color White
Boiling Point 298°C
Quantity 5 g
Formula Weight 208.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material alpha-Asarone
RTECSNumber DC2975000
TSCA No