missing translation for 'onlineSavingsMsg'
Learn More

alpha-Angelicalactone 98.0+%, TCI America™

Catalog No. p-7117238
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A109025G 25 g
A1090100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A109025G Supplier TCI America Supplier No. A109025G
Only null left

Chemical Identifiers

CAS 591-12-8
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.10
MDL Number MFCD00005375
InChI Key QOTQFLOTGBBMEX-UHFFFAOYSA-N
Synonym alpha-angelica lactone, alpha-angelicalactone, 5-methyl-2 3h-furanone, 5-methylfuran-2 3h-one, 2 3h-furanone, 5-methyl, 4-hydroxypent-3-enoic acid lactone, delta 2-angelica lactone, 4-hydroxy-3-pentenoic acid gamma-lactone, gamma-methyl-beta,gamma-crotonolactone, .alpha.-angelica lactone
PubChem CID 11559
ChEBI CHEBI:36433
IUPAC Name 5-methyl-2,3-dihydrofuran-2-one
SMILES CC1=CCC(=O)O1

Specifications

Color Yellow
Boiling Point 56°C
Quantity 25 g
Formula Weight 98.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material alpha-Angelicalactone
EINECSNumber (5)-5891
RTECSNumber LU5075000
TSCA Yes